missing translation for 'onlineSavingsMsg'
Learn More
Learn More
I-Ek Monoclonal Antibody (14-4-4S), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA517763
Code : NEW
Additional Details : Weight : 0.01000kg
Description
MA5-17763 targets I-Ek in FACS applications and shows reactivity with Mouse samples. The MA5-17763 immunogen is c3H.
MHC (major histocompatibility complex) class II molecules are transmembrane glycoproteins expressed on the surface of professional antigen-presenting cells, such as macrophages, dendritic cells and B cells. Before their exposition on the cell surface, the MHC class II molecules react with endocytosed exogenous antigens, which are then presented to the T cells. The antigen-binding grove between MHC class II alpha and beta chain is open at both ends and is 15-24 amino acid residues long. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.Specifications
I-Ek | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
P04230, P14439 | |
H2-Ea-ps, H2-Eb1 | |
C3H | |
50 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Flow Cytometry | |
14-4-4S | |
PE | |
H2-Eb1 | |
AI323765; cell surface glycoprotein; E-alpha-f; H-2 class II histocompatibility antigen, E-D alpha chain; H-2 class II histocompatibility antigen, E-K alpha chain; H-2 class II histocompatibility antigen, E-U alpha chain; H-2 class II histocompatibility antigen, I-E beta chain; H-2Ea; H2-Ea; H2-Ea-ps; H2Eb; H-2Eb; H2-Eb1; H2-IE-alpha; histocompatibility 2, class II antigen E alpha; histocompatibility 2, class II antigen E alpha, pseudogene; histocompatibility 2, class II antigen E beta; Ia3; Ia-3; Ia4; Ia-4; I-E alpha MHC class II; I-E beta MHC class II; I-Ealpha; major histocompatibility complex H-2E beta chain; MGC117888; MHC class II antigen E alpha; MHC class II IE-b; MHC class II protein; MHC H2-IE-beta cell surface glycoprotein | |
Mouse | |
Protein G | |
RUO | |
100504404, 14969 | |
4° C, do not freeze | |
Liquid |