missing translation for 'onlineSavingsMsg'
Learn More

Escherichia coli O157 Mouse anti-E. coli, Clone: 9.88, Abnova™

Product Code. 16043336
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16043336 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16043336 Supplier Abnova Supplier No. MAB15086.100ug
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against Escherichia coli O157.

  • High specificity
  • High reactivity

Specifications

Antigen Escherichia coli O157
Applications Immunofluorescence
Classification Monoclonal
Clone 9.88
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Escherichia coli O157.
Dilution Immunofluorescence (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
Host Species Mouse
Immunogen A crude sonicate of Escherichia coli O157.
Purification Method PEG precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species E. coli
Content And Storage Store at 4°C.
Form Liquid
Isotype IgM κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.