missing translation for 'onlineSavingsMsg'
Learn More

m-Xylene, Fluka™

Product Code. 15959467 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15959467 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 15959467 Supplier Honeywell-Fluka Supplier No. R15835ML

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

Analytical standard, reference material

Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.

  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery
  • Expert Technical Support
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material m-Xylene
Packaging Glass Bottle
CAS 108-38-3
Molecular Formula C6H4(CH3)2
Color Colorless
Melting Point -48°C
Boiling Point 138°C to 139°C (1.013 hPa)
Synonym 1,3-Dimethylbenzene
Molecular Weight (g/mol) 106.17
Quantity 5 mL
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.