missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Silencer™ Human Apoptosis siRNA Library

Product Code. 15651862
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15651862 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15651862 Supplier Invitrogen™ Supplier No. A30096
 Restricted Item
Estimated Shipment: 11-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Silencer™ Human Apoptosis siRNA Library provides investigators with a focused siRNA screening platform targeting apoptosis genes and enabling a detailed analysis of the apoptotic signaling pathway.

The Silencer™ Human Apoptosis siRNA Library provides investigators with a focused siRNA screening platform targeting apoptosis genes and enabling a detailed analysis of the apoptotic signaling pathway. The Silencer Human Apoptosis siRNA Library includes 2769 Ambion™ Silencer siRNAs targeting 923 apoptosis genes for performing RNAi functional studies of apoptotic signaling proteins. The library was designed using the most up-to-date genome databases, including the NCBI RefSeq database and cross-referenced to the Gene Ontology Consortium (GO) database and/or the HUGO Gene Nomenclature Committee (HGNC). Utilizing Ambion Silencer technology built on stringent bioinformatics to enable high target specificity and siRNA potency, the Silencer Human Apoptosis siRNA Library is an ideal solution for focused and cost-effective RNAi screening in apoptosis studies.

  • Comprehensive – Expedite your apoptosis RNAi gene function screens with pre-defined Silencer Human Apoptosis siRNA Library
  • Reliable – Increase target screening accuracy with three individual siRNAs per gene targetEffective – Reduce off target impacts at low concentrations with proven Ambion Silencer siRNA design
  • Convenient – Reduce data de-convolution and accelerate target validation with our arrayed format
  • Customizable – Choose the best option for your workflow demands, including additional siRNA scales and pooled formats

Why use the Silencer Human Apoptosis siRNA Library?
Together with careful bioinformatics selection of gene targets, Silencer siRNAs are optimized with design and high quality synthesis with guaranteed silencing. The Silencer Human Apoptosis siRNA Library contains our pre-designed and when available, validated siRNAs generated with maximum potency and specificity in mind so you can focus on data quality. All Silencer libraries are selected against the latest available databases and resources for up-to-date content. Full siRNA sequence information, as well as the exon targeted and each targeted gene's symbol, full name, aliases, RefSeq number, and Locus ID, plus results from any validation experiments performed using the included siRNAs is provided. Please contact us for the complete list of targets.

Silencer siRNA libraries allow you to screen efficiently and maximize data output
Ambion Silencer siRNAs are designed with potency in mind to help ensure gene knockdown at minimum concentrations. This not only helps ensure less potential for off target effects and false positive data but also minimizes false negative outcomes of siRNA screening. The Silencer Human Apoptosis siRNA Library contains 0.25 nmol of each siRNA, sufficient for 100 transfections (at 25 nM siRNA, 100 μL transfection volume).

Flexibility to meet your workflow needs
Thermo Fisher Scientific offers a comprehensive list of pre-defined or custom siRNA libraries to human, mouse or rat genes with 1, 2, or 5 nmol of each siRNA or larger sizes. For a complete gene list, additional information or to discuss your research with one of our technical experts, contact your local sales representative or e-mail us at RNAilibraries@lifetech.

TRUSTED_SUSTAINABILITY

Specifications

Description 2769 total siRNAs targeting 923 genes plated onto 96-well plates (33 plates)
Content And Storage 2769 total siRNAs targeting 923 genes plated onto 33 x 96-well plates

Store at or below -20°C in a non-frost-free freezer.
Format 96-well Plate
RNAi Type siRNA
Species Human
Sample Type RNA
For Use With (Application) RNAi
Product Line Silencer
Product Type siRNA Library
Purity Standard
Quantity 1 Set
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Show More Show Less
Which Silencer siRNA libraries do you offer?

Our Silencer siRNA library offerings can be found here (https://www.thermofisher.com/us/en/home/life-science/rnai/synthetic-rnai-analysis/sirna-libraries/silencer-sirna-libraries.html).

Which Silencer Select siRNA libraries do you offer?

Our Silencer Select siRNA library offerings can be found here (https://www.thermofisher.com/us/en/home/life-science/rnai/synthetic-rnai-analysis/sirna-libraries/silencer-select-sirna-libraries.html).

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.