missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Silencer™ Select Human Apoptosis siRNA Library

Product Code. 15641862
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15641862 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15641862 Supplier Invitrogen™ Supplier No. A30095
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Silencer™ Select Human Apoptosis siRNA Library provides investigators with a focused siRNA screening platform targeting apoptosis genes and enabling a detailed analysis of the apoptotic signaling pathway.

The Silencer™ Select Human Apoptosis siRNA Library provides investigators with a focused siRNA screening platform targeting apoptosis genes and enabling a detailed analysis of the apoptotic signaling pathway. The Silencer Select Human Apoptosis siRNA Library contains 2772 Ambion™ Silencer Select highly potent and chemically-modified siRNAs targeting 924 apoptosis genes. This library was designed using the most up-to-date genome databases, including the NCBI RefSeq database and cross-referenced to the Gene Ontology Consortium (GO) database and/or the HUGO Gene Nomenclature Committee (HGNC). The Silencer Select siRNAs are designed using Thermo Fisher Scientific's leading design algorithms—along with our novel locked nucleic acid (LNA) chemical modifications, this helps to maximize the efficacy, specificity and potency required for confident RNAi screens during apoptosis investigations.

  • Superior efficacy—Reduce off target effects by up to 90% due to latest design algorithms and LNA chemical modifications
  • Highest efficiency—Perform knockdown studies with 100-fold more potent siRNAs than currently available siRNAs
  • Dependable—Achieve faster discoveries via consistency of gene knockdown and reliability of phenotypic results
  • Convenient—Reduce data de-convolution and accelerate target validation with our arrayed format
  • Customizable—Choose the best option for your workflow demands, including additional siRNA scales and pooled formats

Why use the Silencer Select Human Apoptosis siRNA Library?

Together with careful bioinformatics selection of gene targets, Silencer Select siRNAs are optimized with latest design algorithms, proprietary chemical modifications and high quality synthesis to help ensure desired RNAi outcomes. The Silencer Select Human Apoptosis siRNA Library contains our pre-designed and when available, validated siRNAs generated with highest potency and specificity in mind so you can focus on the highest data quality backed by the best in industry guarantee.

SilencerSelect siRNA libraries empower you with highly potent siRNAs to maximize discovery with even less

Ambion Silencer Select siRNAs are designed with the highest potency of available siRNAs, resulting in 100-fold more potent siRNAs and allowing gene knockdowns at the lowest concentrations (see figure below).The highly potent Silencer Select siRNAs allow you to use even lower concentrations of siRNAs, which helps to minimize off target effects, reduce false negative data and maximize siRNA library output for additional screens and data validation. The Silencer Select Human Apoptosis siRNA Library contains 0.25 nmol of each highly potent siRNA, sufficient for at least 500 transfections (at 5 nM siRNA, 100 μL transfection volume).

Flexibility to meet your workflow needs

Thermo Fisher Scientific offers a comprehensive list of pre-defined or custom siRNA libraries to human, mouse or rat genes with 1, 2, or 5 nmol of each siRNA or larger sizes. For a complete gene list, additional information or to discuss your research with one of our technical experts, contact your local sales representative or e-mail us at RNAilibraries@lifetech.com.

TRUSTED_SUSTAINABILITY

Specifications

Description 2772 total siRNAs targeting 924 genes plated onto 96-well plates (33 plates)
Content And Storage 2772 total siRNAs targeting 924 genes plated onto 33 x 96-well plates

Store at or below -20°C in a non-frost-free freezer.
Format 96-well Plate
RNAi Type siRNA
Species Human
For Use With (Application) RNAi
Product Line Silencer™Select
Product Type siRNA Library
Purity Standard
Quantity 1 Set
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.