missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IAH1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA548275
Additional Details : Weight : 0.01000kg
Description
The function of this protein remains unknown.Specifications
IAH1 | |
Polyclonal | |
Unconjugated | |
Iah1 | |
1500019E20Rik; 4833421E05Rik; Harpb64; hypertrophic agonist responsive protein B64; hypertrophic agonist-responsive protein B64; Iah1; Isoamyl acetate-hydrolyzing esterase 1 homolog; isoamyl acetate-hydrolyzing esterase 1 homolog (S. cerevisiae); testicular tissue protein Li 92 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
285148, 67732 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.35 mg/mL | |
PBS with 0.09% sodium azide | |
Q2TAA2, Q9DB29 | |
Iah1 | |
KLH conjugated synthetic peptide between 78-106 amino acids from the Central region of human IAH1. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |