Learn More
IBA1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10904-1-AP-150UL
 View more versions of this product
Product Code. 16897593
  • 410.00 € / 150µL
Description
AIF1 is induced by cytokines and interferon. It is thought to be involved in the negative regulation of growth of vascular smooth muscle cells, which contributes to the anti-inflammatory response to vessel wall trauma.
Specifications
Specifications
IBA1
Polyclonal
Unconjugated
AIF1
AIF 1, AIF1, G1, IBA1, IRT 1, Protein G1
Rabbit
Antigen Affinity Chromatography
RUO
11629, 199
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
0.31 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O70200, P55008
AIF1
IBA1 Fusion Protein Ag1363
150 μL
Primary
Human, Mouse