Learn More
Invitrogen™ Ibalizumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA559218
 View more versions of this product
Product Code. 30282599
  • 1204.00 € / 1mg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Ibalizumab (also known asibalizumab-uiykand formerly known as TNX-355) is a monoclonal antibody that binds to CD4 receptors on the surface of CD4-positive cells, preventing HIV particle entry into the lymphocytes. It is an advanced and current antibody in development for the treatment of HIV/AIDS. It has been developed by Taimed biologics and Theratechnologies 8, 5.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ibalizumab Humanized
Recombinant Monoclonal
Unconjugated
Hu5A8; TMB-355; TNX-355
Protein A
RUO
Human
Antibody
IgG4SP
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
1 mg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized