missing translation for 'onlineSavingsMsg'
Learn More

IBT6A hydrochloride, MedChemExpress

Product Code. 16491677
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491677 50 mg 50mg
16478217 5 mg 5mg
16436086 100 mg 100mg
16435508 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16491677 Supplier Medchem Express Supplier No. HY13036B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress IBT6A hydrochloride is an impurity of Ibrutinib. IBT6A can be used in synthesis of IBT6A Ibrutinib dimer and IBT6A adduct. Ibrutinib is a selective, irreversible Btk inhibitor with an IC50 of 0.5 nM.

IBT6A hydrochloride is an impurity of Ibrutinib. IBT6A can be used in synthesis of IBT6A Ibrutinib dimer and IBT6A adduct. Ibrutinib is a selective, irreversible Btk inhibitor with an IC50 of 0.5 nM.

Specifications

Chemical Name or Material IBT6A hydrochloride
CAS 1553977-42-6
Color White
Purity Grade Notes Research
Molecular Formula C22H23ClN6O
Quantity 50 mg
Solubility Information DMSO : 100 mg/mL (236.46 mM; Need ultrasonic)
SMILES NC1=C2C(N([C@H]3CNCCC3)N=C2C4=CC=C(OC5=CC=CC=C5)C=C4)=NC=N1.Cl
Molecular Weight (g/mol) 422.91
Formula Weight 422.91
Percent Purity 99.22%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.