missing translation for 'onlineSavingsMsg'
Learn More

IBT6A, MedChemExpress

Product Code. 16426086
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Packungsgröße:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Packungsgröße
16426086 100 mg 100mg
16481677 50 mg 50mg
16468217 5 mg 5mg
16425508 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 16426086 Lieferant Medchem Express Lieferanten-Nr. HY13036A
 Restricted Item
Estimated Shipment: 30-11-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

MedChemExpress IBT6A is an impurity of Ibrutinib. IBT6A can be used in synthesis of IBT6A Ibrutinib dimer and IBT6A adduct. Ibrutinib is a selective, irreversible Btk inhibitor with an IC50 of 0.5 nM.

IBT6A is an impurity of Ibrutinib. IBT6A can be used in synthesis of IBT6A Ibrutinib dimer and IBT6A adduct. Ibrutinib is a selective, irreversible Btk inhibitor with an IC50 of 0.5 nM.

Specifications

Chemical Name or Material IBT6A
CAS 1022150-12-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C22H22N6O
Quantity 100 mg
Solubility Information DMSO : 50 mg/mL (129.38 mM; ultrasonic and warming and heat to 60°C)
SMILES NC1=C2C(N([C@H]3CNCCC3)N=C2C4=CC=C(OC5=CC=CC=C5)C=C4)=NC=N1
Molecular Weight (g/mol) 386.45
Formula Weight 386.45
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.