missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBTK Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00 € - 550.00 €
Specifications
| Antigen | IBTK |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30230247
|
Novus Biologicals
NBP3-35477-20ul |
20 μL |
190.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30232817
|
Novus Biologicals
NBP3-35477-100ul |
100 μL |
550.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IBTK Polyclonal antibody specifically detects IBTK in Human samples. It is validated for ELISA,Western BlotSpecifications
| IBTK | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Bruton agammaglobulinemia tyrosine kinase inhibitor, BTK-binding protein, BTKI, DKFZP564B116, IBtk, inhibitor of Bruton agammaglobulinemia tyrosine kinase, inhibitor of Bruton tyrosine kinase, inhibitor of Bruton's tyrosine kinase, inhibitor of Bruton's tyrosine kinase-alpha, inhibitor of Bruton's tyrosine kinase-beta, inhibitor of Bruton's tyrosine kinase-gamma, KIAA1417, MGC142256, MGC142258, RP1-93K22.1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 900-1200 of human IBTK (NP_056340.2).,, Sequence:, GLNMAALLEARSLDVLSDGVLKDLSEFYRKMIPAMDRRVITPYQDGPDISYLEVEDGDIFLKEEINMEQNHSETMFKKAKTKAKKKPRKRSDSSGGYNLSDIIQSPSSTGLLKSGKTNSVESLPELLTSDSEGSYAGVGSPRDLQSPDFTTGFHSDKIEAKVKPYVNGTSPVYSREDLKPWEKSPILKISAPQPIPSNRIDTTSSASWVAGSFSPVSPPVVDLRTIMEIEESRQKCGATPKSHLGKTVSHGVKLSQKQRKMIALTTKENNSGMNSMETVLFTPSKAPKPVNAWASSLHSVS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 25998 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title