Learn More
IBTK Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA554895
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LKEICEVALT EKLTLKNAAM LLEFAAMYSA KQLKLSCLQF IGLNMAALLE ARSLDVLSDG VLKDLSEFYR KMIPAMDRRV ITPYQDGPDI SYLEVEDGDI FLKEEINMEQ NHSETMFKKA K Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 79%.
The protein encoded by this gene binds to Bruton's tyrosine kinase (BTK) and downregulates BTK's kinase activity. In addition, the encoded protein disrupts BTK-mediated calcium mobilization and negatively regulates the activation of nuclear factor-kappa-B-driven transcription.Specifications
IBTK | |
Polyclonal | |
Unconjugated | |
IBTK | |
Bruton agammaglobulinemia tyrosine kinase inhibitor; BTBD26; BTK-binding protein; BTKI; IBTK; Inhibitor of Bruton tyrosine kinase; inhibitor of Bruton's tyrosine kinase-alpha; inhibitor of Bruton's tyrosine kinase-beta; inhibitor of Bruton's tyrosine kinase-gamma; KIAA1417; RP1-93K22.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25998 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P2D0 | |
IBTK | |
Recombinant protein corresponding to Human IBTK | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only