missing translation for 'onlineSavingsMsg'
Learn More

Ibuprofen, MedChemExpress

Product Code. 16481979
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481979 1 g 1g
16430678 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16481979 Supplier Medchem Express Supplier No. HY78131
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ibuprofen ((±)-Ibuprofen) is a potent, orally active, selective COX-1 inhibitor with an IC50 value of 13 μM. Ibuprofen inhibits cell proliferation, angiogenesis, and induces cell apoptosis. Ibuprofen is a nonsteroidal anti-inflammatory agent and a nitric oxide (NO) donor. Ibuprofen ((±)-Ibuprofen) can be used in the research of pain, swelling, inflammation, infection, immunology, cancers.

Ibuprofen ((±)-Ibuprofen) is a potent, orally active, selective COX-1 inhibitor with an IC50 value of 13 μM. Ibuprofen inhibits cell proliferation, angiogenesis, and induces cell apoptosis. Ibuprofen is a nonsteroidal anti-inflammatory agent and a nitric oxide (NO) donor. Ibuprofen ((±)-Ibuprofen) can be used in the research of pain, swelling, inflammation, infection, immunology, cancers.

Specifications

Chemical Name or Material Ibuprofen
Physical Form Powder
CAS 15687-27-1
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C13H18O2
For Use With (Application) COVID-19-immunoregulation
Synonym (±)-Ibuprofen
Solubility Information DMSO : 100 mg/mL (484.78 mM; Need ultrasonic) ∣H2O : 1 mg/mL (4.85 mM; Need ultrasonic)
SMILES OC(C(C1=CC=C(CC(C)C)C=C1)C)=O
Molecular Weight (g/mol) 206.28
Formula Weight 206.28
Percent Purity 99.5%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.