missing translation for 'onlineSavingsMsg'
Learn More

Ibutilide Fumarate, TRC

Product Code. 30135617
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135617 10 mg 10mg
30134899 5 mg 5mg
30007760 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135617 Supplier Toronto Research Chemicals Supplier No. TRCI15540010MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122647-32-9
Molecular Formula 2 C20 H36 N2 O3 S . C4 H4 O4
Molecular Weight (g/mol) 885.22
Synonym Methanesulfonamide, N-[4-[4-(ethylheptylamino)-1-hydroxybutyl]phenyl]-, (2E)-2-butenedioate (2:1), Methanesulfonamide, N-[4-[4-(ethylheptylamino)-1-hydroxybutyl]phenyl]-, (2E)-2-butenedioate (2:1) (salt) (9CI), Methanesulfonamide, N-[4-[4-(ethylheptylamino)-1-hydroxybutyl]phenyl]-, (±)-, (E)-2-butenedioate (2:1) (salt), Corvert, Ibutilide fumarate, Methanesulfonamide, N-[4-[4-(ethylheptylamino)-1-hydroxybutyl]phenyl]-, (E)-2-butenedioate (2:1) (salt), U 70226E
IUPAC Name (E)-but-2-enedioic acid;N-[4-[4-[ethyl(heptyl)amino]-1-hydroxybutyl]phenyl]methanesulfonamide
SMILES CCCCCCCN(CC)CCCC(O)c1ccc(NS(=O)(=O)C)cc1.CCCCCCCN(CC)CCCC(O)c2ccc(NS(=O)(=O)C)cc2.OC(=O)\C=C\C(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/2C20H36N2O3S.C4H4O4/c2*1-4-6-7-8-9-16-22(5-2)17-10-11-20(23)18-12-14-19(15-13-18)21-26(3,24)25;5-3(6)1-2-4(7)8/h2*12-15,20-21,23H,4-11,16-17H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;;2-1+
Formula Weight 884.5003
Chemical Name or Material Ibutilide Fumarate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.