missing translation for 'onlineSavingsMsg'
Learn More

IC87201, MedChemExpress

Product Code. 16453028
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16453028 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16453028 Supplier Medchem Express Supplier No. HY100457
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress IC87201, an inhibitor of PSD95-nNOS protein-protein interactions, suppresses NMDAR-dependent NO and cGMP formation.

IC87201, an inhibitor of PSD95-nNOS protein-protein interactions, suppresses NMDAR-dependent NO and cGMP formation.

Specifications

Chemical Name or Material IC87201
CAS 866927-10-8
Color Red
Purity Grade Notes Research
Molecular Formula C13H10Cl2N4O
Quantity 10 mg
Solubility Information DMSO : ≥ 28 mg/mL (90.57 mM)
SMILES OC1=C(Cl)C=C(Cl)C=C1CNC2=CC=C3N=NNC3=C2
Molecular Weight (g/mol) 309.15
Formula Weight 309.15
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.