missing translation for 'onlineSavingsMsg'
Learn More

ICA, MedChemExpress

Product Code. 16491909
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491909 1 mg 1mg
16443947 50 mg 50mg
16410488 5 mg 5mg
16408028 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16491909 Supplier Medchem Express Supplier No. HY22044

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress ICA (N-(pyridin-2-yl)-4-(pyridin-2-yl)thiazol-2-amine) is a SK channel inhibitor that has antileishmanial activity with an IC50 of 2.1 μM.

ICA (N-(pyridin-2-yl)-4-(pyridin-2-yl)thiazol-2-amine) is a SK channel inhibitor that has antileishmanial activity with an IC50 of 2.1 μM.

Specifications

Chemical Name or Material ICA
Physical Form Solid
CAS 3374-88-7
Color Brown
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C13H10N4S
For Use With (Application) COVID-19-immunoregulation
Synonym N-[4-(2-Pyridinyl)-2-thiazolyl]-2-pyridinamine
Solubility Information DMSO : ≥ 49.5 mg/mL (194.64 mM)
SMILES C1(NC2=NC(C3=NC=CC=C3)=CS2)=NC=CC=C1
Molecular Weight (g/mol) 254.31
Formula Weight 254.31
Percent Purity 99.63%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.