missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICA1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547573
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
ICA1 Polyclonal antibody specifically detects ICA1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Western Blot.Specifications
ICA1 | |
Polyclonal | |
Unconjugated | |
ICA1 | |
69 kDa islet cell autoantigen; 69kDa; diabetes mellitus type I autoantigen; ica1; ica1 {ECO:0000312; ICA69; ICAp69; Islet cell autoantigen 1; islet cell autoantigen 1 isoform; islet cell autoantigen 1, 69 kDa; islet cell autoantigen 1, 69kDa; islet cell autoantigen p69; p69; RGD:621465}; testicular tissue protein Li 162; Unknown (protein for MGC:134001); zgc:92566 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
3382 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q05084 | |
ICA1 | |
E. coli-derived recombinant human ICA1 Ser2-Ala482 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |