missing translation for 'onlineSavingsMsg'
Learn More

ICAM-1-IN-1, MedChemExpress

Product Code. 16460099
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16460099 10 mg 10mg
16412298 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16460099 Supplier Medchem Express Supplier No. HYU00003
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress ICAM-1-IN-1 is a potent and selective inhibitor of E-selectin and ICAM-1 with IC50 values of 7 and 5 nM, respectively.

ICAM-1-IN-1 is a potent and selective inhibitor of E-selectin and ICAM-1 with IC50 values of 7 and 5 nM, respectively.

Specifications

Chemical Name or Material ICAM-1-IN-1
CAS 251994-14-6
Color White
Purity Grade Notes Research
Molecular Formula C15H11BrN2O2S
Quantity 10 mg
Solubility Information DMSO : ≥ 135 mg/mL (371.67 mM)
SMILES O=C(C(S1)=CC2=C1C=NC=C2OC3=CC=C(Br)C=C3)NC
Molecular Weight (g/mol) 363.23
Formula Weight 363.23
Percent Purity 99.73%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.