missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM-5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-86938-25ul
This item is not returnable.
View return policy
Description
ICAM-5 Polyclonal antibody specifically detects ICAM-5 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| ICAM-5 | |
| Polyclonal | |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| ICAM-5, intercellular adhesion molecule 5, intercellular adhesion molecule 5, telencephalin, TLCNTelencephalin, TLNtelencephalin | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: VRSGELGAVIEGLLRVAREHAGTYRCEATNPRGSAAKNVAVTVEYGPRFEEPSCPSNWTWVEGSGRLFSCEVDGKPQPSVKCVGSGGATEGVLLPLAPPDPSPRAPRIPRVLAPGIYVCNATNRHGSVAKTVVVS | |
| 25 μL | |
| Immunology, Neuroscience | |
| 7087 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction