Learn More
ICAM-5 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547084
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) ICAM-5 and recombinant mouse (rm) ICAM-2 is observed, 5% cross-reactivity with recombinant rat ICAM-1, rhICAM-1, and rhICAM-3 is observed and less than 1% cross-reactivity with rmICAM-1 and rmVCAM-1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
ICAM-5 Polyclonal antibody specifically detects ICAM-5 in Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
ICAM-5 | |
Polyclonal | |
Unconjugated | |
Icam5 | |
CD50; Icam3; ICAM-3; Icam5; ICAM-5; intercellular adhesion molecule 3; intercellular adhesion molecule 5; intercellular adhesion molecule 5, telencephalin; telencephalin; Tlcn; TLN | |
Goat | |
Antigen affinity chromatography | |
RUO | |
15898, 313785 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q60625 | |
Icam5 | |
Mouse myeloma cell line NS0-derived recombinant mouse ICAM-5 Leu31-Arg828 (Pro47Arg) | |
100 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |