missing translation for 'onlineSavingsMsg'
Learn More

Icariin, MedChemExpress

Product Code. 16482336
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16482336 500 mg 500mg
16481476 200 mg 200mg
16458956 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16482336 Supplier Medchem Express Supplier No. HYN0014
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Icariin is a flavonol glycoside. Icariin inhibits PDE5 and PDE4 activities with IC50s of 432 nM and 73.50 μM, respectively. Icariin also is a PPARα activator.

Icariin is a flavonol glycoside. Icariin inhibits PDE5 and PDE4 activities with IC50s of 432 nM and 73.50 μM, respectively. Icariin also is a PPARα activator.

Specifications

Chemical Name or Material Icariin
CAS 489-32-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C33H40O15
Quantity 500 mg
Synonym Ieariline
Solubility Information DMSO : 125 mg/mL (184.73 mM; Need ultrasonic)
SMILES O[C@H]1[C@H](O)[C@@H](CO)O[C@@H](OC2=C(C/C=C(C)/C)C(OC(C3=CC=C(OC)C=C3)=C(O[C@@H]4O[C@@H](C)[C@H](O)[C@@H](O)[C@H]4O)C5=O)=C5C(O)=C2)[C@@H]1O
Molecular Weight (g/mol) 676.66
Formula Weight 676.66
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.