missing translation for 'onlineSavingsMsg'
Learn More

ICI 118,551 hydrochloride, MedChemExpress

Product Code. 16476366
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16476366

Brand: Medchem Express HY13951

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ICI 118,551 (hydrochloride) is a highly selective β2 adrenergic receptor antagonist, with Kis of 0.7, 49.5 and 611 nM for β2, β1 and β3 receptors, respectively.

ICI 118,551 (hydrochloride) is a highly selective β2 adrenergic receptor antagonist, with Kis of 0.7, 49.5 and 611 nM for β2, β1 and β3 receptors, respectively.

Specifications

Chemical Name or Material ICI 118,551 hydrochloride
CAS 72795-01-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H28ClNO2
Quantity 100 mg
Synonym ICI 118551 hydrochloride
Solubility Information DMSO : 33.33 mg/mL (106.19 mM; Need ultrasonic) ∣H2O : 12.5 mg/mL (39.83 mM; Need ultrasonic)
SMILES CC1=C(CCC2)C2=C(OC[C@H](O)[C@@H](C)NC(C)C)C=C1.[H]Cl
Molecular Weight (g/mol) 313.86
Formula Weight 313.86
Percent Purity 97.95%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.