missing translation for 'onlineSavingsMsg'
Learn More

ICI 199,441 hydrochloride, Tocris Bioscience™
GREENER_CHOICE

Product Code. 30001129
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001129

Brand: Tocris Bioscience 0778/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Potent κ agonist

Highly potent κ agonist, 146-fold more active than U-50488 (Cat. No. 0495) in vitro.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115199-84-3
Molecular Formula C21H25Cl3N2O
Molecular Weight (g/mol) 427.794
InChI Key VFLWVWZSDBTGQJ-VEIFNGETSA-N
Synonym ici-199,441 hydrochloride, ici 199,441 hydrochloride, n-2-n-methyl-3,4-dichlorophenylacetamido-2-phenylethyl pyrrolidine, s-3,4-dichloro-n-methyl-n-1-phenyl-2-1-pyrrolidinyl ethyl benzeneacetamide monohydrochloride, 2-3,4-dichlorophenyl-n-methyl-n-1s-1-phenyl-2-1-pyrrolidinyl ethyl acetamide hydrochloride, 2-3,4-dichlorophenyl-n-methyl-n-1s-1-phenyl-2-pyrrolidin-1-yl ethyl acetamide hydrochloride, 2-3,4-dichlorophenyl-n-methyl-n-1s-1-phenyl-2-pyrrolidin-1-ylethyl acetamide hydrochloride, benzeneacetamide, 3,4-dichloro-n-methyl-n-1-phenyl-2-1-pyrrolidinyl ethyl-, monohydrochloride, s
PubChem CID 3082717
IUPAC Name 2-(3,4-dichlorophenyl)-N-methyl-N-[(1S)-1-phenyl-2-pyrrolidin-1-ylethyl]acetamide;hydrochloride
SMILES CN(C(CN1CCCC1)C2=CC=CC=C2)C(=O)CC3=CC(=C(C=C3)Cl)Cl.Cl

Specifications

Quantity 10 mg
Formula Weight 427.8
Percent Purity >99%
Chemical Name or Material ICI 199, 441 hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.