missing translation for 'onlineSavingsMsg'
Learn More

ICP-AES CRQL Solution 1: Be, Cd @ 5; Ag, Cr, Pb @ 10; As, Mn @ 15; Cu, Tl @ 25; Se @ 35; Ni @ 40; Co, V @ 50; Sb, Zn @ 60; Fe @ 100; Al, Ba @ 200 μg/mL in 5% HNO3, tr. Tartaric Acid, VHG™

Product Code. 30003187
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30003187 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 30003187 Supplier VHG Labs Supplier No. VHGCRQL1AES100
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

ICP-AES CRQL Solution 1: Be, Cd @ 5; Ag, Cr, Pb @ 10; As, Mn @ 15; Cu, Tl @ 25; Se @ 35; Ni @ 40; Co, V @ 50; Sb, Zn @ 60; Fe @ 100; Al, Ba @ 200 μg/mL in 5% HNO3, tr. Tartaric Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Beryllium/Cadmium/Chromium/Lead/Silver/Arsenic/Manganese/Copper/Thallium/Selenium/Nickel/Cobalt/Vanadium/Antimony/Zinc/Iron/Aluminium/Barium
CAS 7440-41-7, 7440-43-9, 7440-47-3, 7439-92-1, 7440-22-4, 7440-38-2
Methods ICP
Analyte or Component Names Beryllium/Cadmium/Chromium/Lead/Silver/Arsenic/Manganese/Copper/Thallium/Selenium/Nickel/Cobalt/Vanadium/Antimony/Zinc/Iron/Aluminium/Barium
Shipping Condition Room Temperature
Solvent or Matrix Dilute HNO3
Quantity 100 mL
Concentration or Composition (by Analyte or Components) Beryllium 5 μg/mL/Cadmium 5 μg/mL/Chromium 10 μg/mL/Lead 10 μg/mL/Silver 10 μg/mL/Arsenic 15 μg/mL/Manganese 15 μg/mL/Copper 25 μg/mL/Thallium 25 μg/mL/Selenium 35 μg/mL/Nickel 40 μg/mL/Cobalt 50 μg/mL/Vanadium 50 μg/mL/Antimony 60 μg/mL/Zinc 60 μg/mL/Iron 100 μg/mL/Aluminium 200 μg/mL/Barium 200 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.