missing translation for 'onlineSavingsMsg'
Learn More

ICP, ICP-MS Multi-Element Standard (19 Elements) in 5% Nitric Acid (HNO3), Reagecon
BIOPHARMA

Product Code. 11419079 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11419079 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11419079 Supplier Reagecon Supplier No. ICP19A10
  • ONLINE EXCLUSIVE PRICE
    609.00 €
    459.54 € / 100mL
    Save 149.46 €
    24% Off
View all promotions
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Manufactured from ultra-pure metals or salts assayed by titration and ICP-MS prior to manufacture

  • ISO/IEC 17025 for the testing of ICP/ICP-MS Standards using ICP-MS (INAB Ref: 264T)
  • ISO/IEC 17025 for calibration of laboratory balances (INAB Ref: 265C)
  • ISO/IEC 17025 for temperature calibration in the range of -196 - +1200°C (INAB Ref: 265C)
  • ISO/IEC 17025 for density measurement (INAB Ref: 264T) using Oscillating U-Tube and Bingham Pycnometry
  • ISO/IEC 17025 for certain complexometric titrations (INAB Ref: 264T)
  • GHS/OSHA compliant, REACH etc.
  • This product is produced in a highly controlled cleanroom (ISO 7) environment, using ultra-pure water (specially treated for the production of mass spectroscopy standards), triple distilled acids and triple-leached pre-cleaned bottles.
  • The titrimetry and spectroscopy are controlled and calibrated using separate Certified Reference Materials (CRM’s), so the product comes with two layers of traceability including NIST Standards, where available.
  • All of the uncertainties of measurement for this product are calculated according to Eurachem/CITAC guidelines and reported as expanded uncertainties at the 95% confidence level.
  • Multi-element standard (19 elements):
  • DescriptionConcentration
    Aluminum (Al)100 μg/mL (ppm)
    Barium (Ba)5 μg/mL (ppm)
    Beryllium (Be)1 μg/mL (ppm)
    Bismuth (Bi)200 μg/mL (ppm)
    Boron (B)15 μg/mL (ppm)
    Cadmium (Cd)20 μg/mL (ppm)
    Chromium (Cr)25 μg/mL (ppm)
    Cobalt (Co)20 μg/mL (ppm)
    Copper (Cu)30 μg/mL (ppm)
    Gallium (Ga)150 μg/mL (ppm)
    Indium (In)200 μg/mL (ppm)
    Iron (Fe)15 μg/mL (ppm)
    Lead (Pb)200 μg/mL (ppm)
    Manganese (Mn)5 μg/mL (ppm)
    Nickel (Ni)50 μg/mL (ppm)
    Silver (Ag)50 μg/mL (ppm)
    Strontium (Sr)1 μg/mL (ppm)
    Thallium (Tl)40 μg/mL (ppm)
    Zinc (Zn)20 μg/mL (ppm)

Specifications

Description Matrix: 5% HNO₃
Packaging HDPE Triple-leached
Analyte or Component Names Multi-element
Physical Form Liquid
Concentration Mixed concentrations of each element
Technique ICP, ICP-MS, AA, Ion Chromatography, XRF, Cation Measurement
Element Multi-element
Quantity 100 mL
compliance-icons compliance-icons
Product Identifier
  • MULTIELEMENT ICP STANDARD
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: nitric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.