missing translation for 'onlineSavingsMsg'
Learn More

ICP, ICP-MS Multi Element Standard in 2 to 5% Nitric Acid (15 Elements), Reagecon
BIOPHARMA

Product Code. 11469079 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11469079 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Artikelnummer. 11469079 Leverandør Reagecon Leverandørnr. ICP15A10
  • ONLINE EXCLUSIVE PRICE
    920.00 €
    459.54 € / 100mL
    Save 460.46 €
    50% Off
Se alle kampagner
 Restricted Item
Estimated Shipment: 19-01-2027
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.

Manufactued from ultra-pure metals or salts

  • Raw materials used are assayed by titration and ICP-MS
  • Separate standards used to control or calibrate titration/ICP-MS
  • Manufactured gravimetrically using mass balance
    • 100% less sum of all impurities
  • ISO/IEC 17025
    • For testing of ICP/ICP-MS standards using ICP-MS
    • For calibration of laboratory balances
    • For temperature calibration -90 to 650 °C
    • For density measurement using Oscillating U-tube and Bingham pycnometry
    • For certain complexometric titrations
  • GHS/OSHA compliant, REACH
  • Produced in highly controlled cleanroom environment using ultra-pure water, triple distilled acids and triple leached pre-cleaned bottles
  • All uncertainties of measurement calculated according to Eurachem/CITAC guidelines and reported at 95% confidence level

Tekniske data

Packaging HDPE triple-leached
Analyte or Component Names 100 μg/mL each of aluminum, barium, calcium cadmium, cobalt, chromium, copper, iron, magnesium, manganese, sodium, nickel, lead, titanium, zinc
Physical Form Liquid
Technique ICP, ICP-MS, AA, Ion Chromatography, XRF, Cation Measurement
UN Number 1760
Type ICP/ICP-MS Standard
Element Al, Ba, Ca, Cd, Co, Cr, Cu, Fe, Mg, Mn, Na, Ni, Pb, Ti, Zn
Solvent or Matrix 2 to 5% HNO3
Quantity 100 mL
compliance-icons compliance-icons
Produktidentifikator
  • MULTIELEMENT ICP STANDARD
Signalord
  • Danger
Farekategori
  • Carcinogenicity Category 1A
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Faresætning
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H350-May cause cancer.
Sikkerhedssætning
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplerende oplysninger
  • MIXTURE LIST-Contain: cobalt carbonate, nitric acid
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.