missing translation for 'onlineSavingsMsg'
Learn More

ICP-MS CRQL Solution 2: Ag, As, Be, Cd, Co, Mn, Ni, Pb, Tl, V @ 10; Cr, Cu, Sb, Zn @ 20; Se @ 50; Ba @ 100 μg/mL in 5% HNO3, tr. Tartaric Acid, VHG™

Product Code. 17918709
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17918709 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 17918709 Supplier VHG Labs Supplier No. VHGCRQL2MS100
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

ICP-MS CRQL Solution 2: Ag, As, Be, Cd, Co, Mn, Ni, Pb, Tl, V @ 10; Cr, Cu, Sb, Zn @ 20; Se @ 50; Ba @ 100 μg/mL in 5% HNO3, tr. Tartaric Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Arsenic/Beryllium/Cadmium/Cobalt/Lead/Manganese/Nickel/Silver/Thallium/Vanadium/Antimony/Chromium/Copper/Zinc/Selenium/Barium
CAS 7440-38-2, 7440-41-7, 7440-43-9, 7440-48-4, 7439-92-1, 7439-96-5
Methods ICP-MS
Analyte or Component Names Arsenic/Beryllium/Cadmium/Cobalt/Lead/Manganese/Nickel/Silver/Thallium/Vanadium/Antimony/Chromium/Copper/Zinc/Selenium/Barium
Shipping Condition Room Temperature
Solvent or Matrix Dilute HNO3
Quantity 100 mL
Concentration or Composition (by Analyte or Components) Arsenic 10 μg/mL/Beryllium 10 μg/mL/Cadmium 10 μg/mL/Cobalt 10 μg/mL/Lead 10 μg/mL/Manganese 10 μg/mL/Nickel 10 μg/mL/Silver 10 μg/mL/Thallium 10 μg/mL/Vanadium 10 μg/mL/Antimony 20 μg/mL/Chromium 20 μg/mL/Copper 20 μg/mL/Zinc 20 μg/mL/Selenium 50 μg/mL/Barium 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.