missing translation for 'onlineSavingsMsg'
Learn More

ICP-OES ICAL Solution: S @ 50 μg/mL; Ce, Cu, Eu, Fe, In, K, Ni, P, Si, Ti, V, Y, Zr @ 10μg/mL; Mn, Mo, Na, Sc @ 5μg/mL; Be, Li, Sr @ 2μg/mL; Ca @ 1 μg/mL in 2% HNO3 / 2% HCl, VHG™

Product Code. 30003154
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30003154

Brand: VHG Labs VHGISUSPCTICAL250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

ICP-OES ICAL Solution: S @ 50 μg/mL; Ce, Cu, Eu, Fe, In, K, Ni, P, Si, Ti, V, Y, Zr @ 10μg/mL; Mn, Mo, Na, Sc @ 5μg/mL; Be, Li, Sr @ 2μg/mL; Ca @ 1 μg/mL in 2% HNO3 / 2% HCl
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Calcium/Beryllium/Lithium/Strontium/Manganese/Molybdenum/Scandium/Sodium/Cerium/Copper/Europium/Indium/Iron/Nickel/Phosphorus/Potassium/Silicon/Titanium/Vanadium/Yttrium/Zirconium
CAS 7440-70-2, 7440-41-7, 7439-93-2, 7440-24-6, 7439-96-5, 7439-98-7
Analyte or Component Names Calcium/Beryllium/Lithium/Strontium/Manganese/Molybdenum/Scandium/Sodium/Cerium/Copper/Europium/Indium/Iron/Nickel/Phosphorus/Potassium/Silicon/Titanium/Vanadium/Yttrium/Zirconium
Shipping Condition Room Temperature
Solvent or Matrix Dilute HNO3
Quantity 250 mL
Concentration or Composition (by Analyte or Components) Calcium 1 μg/mL/Beryllium 2 μg/mL/Lithium 2 μg/mL/Strontium 2 μg/mL/Manganese 5 μg/mL/Molybdenum 5 μg/mL/Scandium 5 μg/mL/Sodium 5 μg/mL/Cerium 10 μg/mL/Copper 10 μg/mL/Europium 10 μg/mL/Indium 10 μg/mL/Iron 10 μg/mL/Nickel 10 μg/mL/Phosphorus 10 μg/mL/Potassium 10 μg/mL/Silicon 10 μg/mL/Titanium 10 μg/mL/Vanadium 10 μg/mL/Yttrium 10 μg/mL/Zirconium 10 μg/mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.