missing translation for 'onlineSavingsMsg'
Learn More

ICP-OES Instrument Check Standard 3: K, P, S @ 100 μg/mL; As, La, Li, Mn, Mo, Na, Ni, Sc @ 20 μg/mL in 5% HCl, VHG™

Product Code. 30003099
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003099 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30003099 Supplier VHG Labs Supplier No. VHGISUPECHKSTD3250

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

ICP-OES Instrument Check Standard 3: K, P, S @ 100 μg/mL; As, La, Li, Mn, Mo, Na, Ni, Sc @ 20 μg/mL in 5% HCl
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Arsenic/Lanthanum/Lithium/Manganese/Molybdenum/Nickel/Scandium/Sodium/Phosphorus/Potassium
CAS 7440-38-2, 7439-91-0, 7439-93-2, 7439-96-5, 7439-98-7, 7440-02-0
Analyte or Component Names Arsenic/Lanthanum/Lithium/Manganese/Molybdenum/Nickel/Scandium/Sodium/Phosphorus/Potassium
Shipping Condition Room Temperature
Solvent or Matrix Dilute HCl
Quantity 250 mL
Concentration or Composition (by Analyte or Components) Arsenic 20 μg/mL/Lanthanum 20 μg/mL/Lithium 20 μg/mL/Manganese 20 μg/mL/Molybdenum 20 μg/mL/Nickel 20 μg/mL/Scandium 20 μg/mL/Sodium 20 μg/mL/Phosphorus 100 μg/mL/Potassium 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.