Learn More
ID1 Antibody (CL11981) [mFluor Violet 500 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-21206MFV500
Product Code. 30294973
  • 759.00 € / 0.10mL
Description
ID1 Monoclonal antibody specifically detects ID1 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ID1
Monoclonal
mFluor Violet 500 SE
BHLHB24, bHLHb24DNA-binding protein inhibitor ID-1, Class B basic helix-loop-helix protein 24, dJ857M17.1.2, dJ857M17.1.2 (inhibitor of DNA binding 1, dominant negative helix-loop-helixprotein), ID, inhibitor of differentiation 1, Inhibitor of DNA binding 1, inhibitor of DNA binding 1, dominant negative helix-loop-helix protein
This antibody was developed using a synthetic peptide derived from P41134, with the exact immunogen sequence remaining proprietary.
0.1 mL
Stem Cell Markers
3397
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL11981
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified