Learn More
ID2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528939
Code : A0
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: HCT116. Predicted reactivity: Mouse (98%), Rat (97%), Xenopus laevis (82%), Pig (98%), Chicken (94%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The inhibitor of DNA binding 2 (ID2) protein is essential for promoting the development of a number of cell types including natural killer cell, erythroid precursors, as well as, CD8+ and CD4+ T-cell. A lack of ID2 promotes B-cell development. The chief role of this protein is to associate with ubiquitously-expressed E proteins, and other proteins of the basic helix-loop-helix (HLH) family, preventing them from binding to DNA and activating transcription. Activation of ID2 is critical for the development of the natural killer cell lineage.Specifications
ID2 | |
Polyclonal | |
Unconjugated | |
ID2 | |
Ac2-300; AI255428; bHLHb26; C78922; cell growth-inhibiting gene 8; class B basic helix-loop-helix protein 26; DNA-binding protein inhibitor ID2; DNA-binding protein inhibitor ID-2; GIG8; helix-loop-helix protein ID2; Id2; Id-2; ID2A; ID2H; Idb2; Inhibitor of differentiation 2; inhibitor of DNA binding 2; inhibitor of DNA binding 2, dominant negative helix-loop-helix protein; inhibitor of DNA binding 2, HLH protein; MGC26389; OTTHUMP00000140258; OTTHUMP00000200297 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
15902, 25587, 3398 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
0.82 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P41136, P41137, Q02363 | |
ID2 | |
Full length human ID2 Recombinant protein. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |