missing translation for 'onlineSavingsMsg'
Learn More

Idarubicin hydrochloride, MedChemExpress

Product Code. 16493327
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16493327 50 mg 50mg
16457318 10 mg 10mg
16409827 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16493327 Supplier Medchem Express Supplier No. HY17381
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Idarubicin hydrochloride is an anthracycline antileukemic drug. It inhibits the topoisomerase II interfering with the replication of DNA and RNA transcription. Idarubicin hydrochloride inhibits the growth of bacteria and yeasts.

Idarubicin hydrochloride is an anthracycline antileukemic drug. It inhibits the topoisomerase II interfering with the replication of DNA and RNA transcription. Idarubicin hydrochloride inhibits the growth of bacteria and yeasts.

Specifications

Chemical Name or Material Idarubicin hydrochloride
CAS 57852-57-0
Color Orange
Purity Grade Notes Research
Molecular Formula C26H28ClNO9
Quantity 50 mg
Synonym 4-Demethoxydaunorubicin hydrochloride
Solubility Information DMSO : 83.33 mg/mL (156.06 mM; Need ultrasonic)
SMILES OC(C(C(C1=CC=CC=C21)=O)=C3C2=O)=C4[C@H](C[C@@](C(C)=O)(O)CC4=C3O)O[C@@](O[C@@H](C)[C@H]5O)([H])C[C@@H]5N.Cl
Molecular Weight (g/mol) 533.95
Formula Weight 533.95
Percent Purity 98.06%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.