missing translation for 'onlineSavingsMsg'
Learn More

IDE1, MedChemExpress

Product Code. 16465807
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16465807 5 mg 5mg
16462606 25 mg 25mg
16450669 1 mg 1mg
16439636 50 mg 50mg
16413078 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16465807 Supplier Medchem Express Supplier No. HY100533
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress IDE1 is an inducer of definitive endoderm 1.

IDE1 is an inducer of definitive endoderm 1.

Specifications

Chemical Name or Material IDE1
Physical Form Solid
CAS 1160927-48-9
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C15H18N2O5
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 30 mg/mL (97.94 mM)
SMILES OC(C1=C(/C=N/NC(CCCCCC(O)=O)=O)C=CC=C1)=O
Molecular Weight (g/mol) 306.31
Formula Weight 306.31
Percent Purity 98.03%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.