missing translation for 'onlineSavingsMsg'
Learn More

Idelalisib, MedChemExpress

Product Code. 16481667
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16481667 50 mg 50mg
16425498 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16481667 Supplier Medchem Express Supplier No. HY13026
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Idelalisib (CAL-101; GS-1101) is a highly selective and orally bioavailable p110δ inhibitor with an IC50 of 2.5 nM, showing 40- to 300-fold selectivity for p110δ over other PI3K class I enzymes.

Idelalisib (CAL-101

Specifications

Chemical Name or Material Idelalisib
CAS 870281-82-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H18FN7O
Quantity 50 mg
Synonym CAL-101 GS-1101
Solubility Information DMSO : ≥ 59.7 mg/mL (143.71 mM)
SMILES O=C1N(C2=CC=CC=C2)C([C@@H](NC3=C4N=CNC4=NC=N3)CC)=NC5=C1C(F)=CC=C5
Molecular Weight (g/mol) 415.42
Formula Weight 415.42
Percent Purity 95.07%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.