Learn More
IFI30 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555365
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LVPYGNAQEQ NVSGRWEFKC QHGEEECKFN KVEACVLDEL DMELAFLTIV CMEEFEDMER SLPLCLQLYA PGLSPDTIME CAMGDRGMQL MHANAQRTDA LQPPHEYVPW VTVNGKPLED QTQLLTLVCQ LYQGKK Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 74%.
The protein encoded by this gene is a lysosomal thiol reductase that at low pH can reduce protein disulfide bonds. The enzyme is expressed constitutively in antigen-presenting cells and induced by gamma-interferon in other cell types. This enzyme has an important role in MHC class II-restricted antigen processing.Specifications
IFI30 | |
Polyclonal | |
Unconjugated | |
IFI30 | |
Gamma-interferon-inducible lysosomal thiol reductase; gamma-interferon-inducible protein IP-30; GILT; IFI30; IFI-30; IFI30, lysosomal thiol reductase; interferon gamma inducible protein 30; interferon gamma-inducible protein 30 preproprotein; interferon, gamma-inducible protein 30; IP30; IP-30; Legumaturain | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10437 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P13284 | |
IFI30 | |
Recombinant protein corresponding to Human IFI30 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only