missing translation for 'onlineSavingsMsg'
Learn More

IFIH1 Mouse anti-Human, Clone: OTI1C7, lyophilized, TrueMAB™

Product Code. 17607621
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17607621 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17607621 Supplier Origene Technologies Supplier No. CF803665
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

Melanoma differentiation associated gene 5, MDA5, is a pattern recognition receptor to transmit the signal.Once activated, both proteins signal through IPS-1 activating transcription factors NF- kappaB and IRF-3 and ultimately activating apoptosis, cytokine signaling, and inflammation. RIG-I is essential for signaling by influenza A, influenza B, human respiratory syncytial virus, paromyxoviruses, Japanese encephalitis virus. MDA5 is essential for signaling by picornavirus. Both RIG-I and MDA5 are responsible for signaling by West Nile virus.

Specifications

Antigen IFIH1
Applications Western Blot
Classification Monoclonal
Clone OTI1C7
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene IFIH1
Gene Accession No. Q9BYX4
Gene Alias AGS7, Hlcd, IDDM19, MDA-5, MDA5, RLR-2, SGMRT1
Gene Symbols IFIH1
Host Species Mouse
Immunogen Full length human recombit protein of human IFIH1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64135
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.