missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFITM1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98695-50ul
This item is not returnable.
View return policy
Description
IFITM1 Polyclonal antibody specifically detects IFITM1 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| IFITM1 | |
| Polyclonal | |
| Western Blot 1:500-1:1000, Immunoprecipitation 0.2-1 μL/mg of lysate, Immunohistochemistry-Paraffin 1:10000-1:50000 | |
| CD225 antigen, CD225Leu-13 antigen, IFI17LEU13,9-27, interferon induced transmembrane protein 1 (9-27), Interferon-induced protein 17, interferon-induced transmembrane protein 1, Interferon-inducible protein 9-27 | |
| Produced in rabbits immunized with A synthetic peptide corresponding to the N-terminus of the human IFITM1. | |
| 50 μg | |
| Cell Cycle and Replication | |
| 8519 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction