missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFITM3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA511274
Code : LE
Additional Details : Weight : 0.01000kg
Description
IFITM3 (interferon induced transmembrane protein 3), also known as 1-8U or IP15, is a multi-pass membrane protein that belongs to the IFITM (interferon inducible transmembrane) family of proteins. IFITM proteins are induced by type I and type II interferons and contain multiple interferon (IFN)-stimulated response elements (ISREs) in their promoter regions. IFITM proteins play important roles in many cellular processes and their expression requires the presence of the chromatin remodeling SWI/SNF-like BAF complexes. Cellular processes involving IFITM proteins include cellular anti-proliferative activities and homotypic cell adhesion functions of interferons. In addition, IFITM genes are often upregulated in various cancer cells, suggesting a possible role in carcinogenesis. Localizing to the membrane, IFITM3 is a 133 amino acid protein that is induced by IFN-alpha and IFN-gamma. IFITM3 expression can be regulated by TEF-1, Brg-1 and Sp1.Specifications
IFITM3 | |
Polyclonal | |
Unconjugated | |
IFITM3 | |
1110004C05Rik; 1-8U; Cd225; Cdw217; Dispanin subfamily A member 2b; DSPA2b; Fgls; Fragilis; Fragilis 3; Fragilis protein; Ifitm3; ifitm-like protein 1; interferon induced transmembrane protein 3; interferon-induced transmembrane protein 3; interferon-inducible protein 15; Interferon-inducible protein 1-8U; interferon-inducible protein variant 10; IP15; mil-1; Mouse ifitm-like protein 1; rat8 | |
Rabbit | |
Affinity chromatography, Protein A | |
RUO | |
10410, 361673, 66141 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Western Blot | |
0.50 mg/mL | |
PBS with 0.09% sodium azide | |
P26376, Q01628, Q9CQW9 | |
IFITM3 | |
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human Interferon-inducible protein (IFITM3) | |
400 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |