missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IFITM3 Recombinant Rabbit Monoclonal Antibody (24GB5155)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557043
This item is not returnable.
View return policy
Description
IFITM3 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
IFITM3 (interferon induced transmembrane protein 3), also known as 1-8U or IP15, is a multi-pass membrane protein that belongs to the IFITM (interferon inducible transmembrane) family of proteins. IFITM proteins are induced by type I and type II interferons and contain multiple interferon (IFN)-stimulated response elements (ISREs) in their promoter regions. IFITM proteins play important roles in many cellular processes and their expression requires the presence of the chromatin remodeling SWI/SNF-like BAF complexes. Cellular processes involving IFITM proteins include cellular anti-proliferative activities and homotypic cell adhesion functions of interferons. In addition, IFITM genes are often upregulated in various cancer cells, suggesting a possible role in carcinogenesis. Localizing to the membrane, IFITM3 is a 133 amino acid protein that is induced by IFN-alpha and IFN-gamma. IFITM3 expression can be regulated by TEF-1, Brg-1 and Sp1.
Specifications
| IFITM3 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| IFITM3 | |
| 1110004C05Rik; 1-8U; Cd225; Cdw217; Dispanin subfamily A member 2b; DSPA2b; Fgls; Fragilis; Fragilis 3; Fragilis protein; Ifitm3; ifitm-like protein 1; interferon induced transmembrane protein 3; interferon-induced transmembrane protein 3; interferon-inducible protein 15; Interferon-inducible protein 1-8U; interferon-inducible protein variant 10; IP15; mil-1; Mouse ifitm-like protein 1; rat8 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10410 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB5155 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q01628 | |
| IFITM3 | |
| A synthesized peptide derived from human Fragilis (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction