missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFITM5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA558715
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SIKARDQKVV GDLEAARRFG SKAKC Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 76%.
Plays a role in bone mineralization.Specifications
IFITM5 | |
Polyclonal | |
Unconjugated | |
IFITM5 | |
1110003J06Rik; AW213665; bone-restricted ifitm-like protein; bone-restricted interferon induced transmembrane protein-like protein; bone-restricted interferon-induced transmembrane protein-like protein; Bril; dispanin subfamily A member 1; DSPA1; Fragilis family member 4; fragilis4; haemopoiesis related membrane protein 1; Hrmp1; Hrtm1; Ifitm5; interferon induced transmembrane protein 5; interferon-induced transmembrane protein 5; OI5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
387733 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6NNB3 | |
IFITM5 | |
Recombinant protein corresponding to Human IFITM5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only