Learn More
IFN epsilon Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA546164
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: IFSLFRANIS LDGWEENHTE KFLIQLHQQL EYLEALMGLE AEKLSGTLGS Sequence homology: Cow: 93%; Human: 100%; Pig: 79% PA5-46164 was successfully used to detect IFNE in endometrial tissue sections by IHC.
IFN epsilon Polyclonal antibody specifically detects IFN epsilon in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
IFN epsilon | |
Polyclonal | |
Unconjugated | |
IFNE | |
IFNE; IFN-E; Ifne1; IFN-epsilon; Ifnt1; Ifn-tau-1; Infe1; Interferon epsilon; interferon epsilon 1; interferon epsilon-1; Interferon tau-1; interferon, epsilon; interferon-epsilon; PRO655; UNQ360/PRO655 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
338376 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q86WN2 | |
IFNE | |
Synthetic peptide directed towards the middle region of human IFNE | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |