missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-gamma Antibody (IFNG/466), Novus Biologicals™
Mouse Monoclonal Antibody
290.00 € - 555.00 €
Specifications
| Antigen | IFN-gamma |
|---|---|
| Clone | IFNG/466 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml, SDS-Page |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18230281
|
Novus Biologicals
NBP2-53332-20UG |
20 μg |
290.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18796743
|
Novus Biologicals
NBP2-53332-100UG |
555.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
IFN-gamma Monoclonal specifically detects IFN-gamma in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| IFN-gamma | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page | |
| Unconjugated | |
| Mouse | |
| Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity, Stem Cell Markers | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| 3458 | |
| Purified recombinant human IFN-gamma | |
| Primary | |
| Store at 4C. |
| IFNG/466 | |
| Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml, SDS-Page | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
| IFNG | |
| IgG2b κ | |
| Protein A or G purified | |
| 22 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title