Learn More
IFN gamma Rat, Biotin, Clone: XMG1.2, enQuireBio™
Rat Monoclonal Antibody
Brand:  EnquireBio QAB83-B-25ug
 View more versions of this product
Product Code. 15982548
  • 116.55 € / 25µg
Specifications
IFN gamma
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2
Ifng
Protein A/G Purified
RUO
15978
IgG1 κ
Flow Cytometry
XMG1.2
Biotin
NP_032363
Rat
25 μg
Primary
Store at 2-4°C.