Learn More
IFN gamma Rat, PE-Cy7, Clone: XMG1.2, enQuireBio™
Rat Monoclonal Antibody
Brand:  EnquireBio QAB83-PE7-100ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15932558
  • 305.00 € / 100µg
Estimated Shipment: 30-01-2023
to see stock.
Specifications
IFN gamma
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2
Ifng
Protein A/G Purified
RUO
15978
IgG1 κ
Flow Cytometry
XMG1.2
PE-Cyanine7
NP_032363
Rat
100 μg
Primary
Store at 2-4°C.