missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFNAR2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10522-1-AP-150UL
This item is not returnable.
View return policy
Description
The type I interferons, IFN-alpha and IFN-beta, are a group of structurally and functionally related proteins that are induced by either viruses or double-stranded RNA and are defined by their ability to confer an antiviral state in cells. IFN-alpha and IFN-beta appear to compete with one another for binding to a common cell surface receptor, while immune IFN (IFN-gamma) binds to a distinct receptor. This distinct receptor, IFN-alphaR, is only weakly responsive to type I interferons, in contrast to IFN-alpha/betaR, which binds to and responds effectively to IFN-beta and to several of the IFN-alpha subtypes. IFN-alpha/betaR is expressed as two alternatively spliced transcripts, designated IFN-alpha/betaRalpha (IFN-alpha/betaR1) and IFN-alpha/betaRbeta (IFN-alpha/betaR2), both of which are involved in signal transduction and ligand binding.Specifications
| IFNAR2 | |
| Polyclonal | |
| Unconjugated | |
| IFNAR2 | |
| IFN alpha REC, IFN alpha/beta receptor 2, IFN R, IFN R 2, IFNABR, IFNAR2, IFNARB, Type I interferon receptor 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3455 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P48551 | |
| IFNAR2 | |
| IFNAR2 Fusion Protein Ag0191 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction