Learn More
IFNK Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA545332
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: MKPSEARVPQ LSSLELRRYF HRIDNFLKEK KYSDCAWEIV RVEIRRCLYY Sequence homology: Cow: 85%; Dog: 93%; Goat: 77%; Guinea Pig: 85%; Horse: 93%; Human: 100%; Pig: 92%; Rat: 77%; Sheep: 85%
IFNK Polyclonal antibody specifically detects IFNK in Human samples. It is validated for Western Blot.Specifications
IFNK | |
Polyclonal | |
Unconjugated | |
IFNK | |
Ifnk; IFN-kappa; IFNT1; INFE1; interferon; Interferon kappa; interferon, kappa; interferon-like protein; RP11-27J8.1; type I interferon kappa; UNQ6124/PRO20084 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
56832 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9P0W0 | |
IFNK | |
Synthetic peptide directed towards the C-terminal of human IFNK | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |