missing translation for 'onlineSavingsMsg'
Learn More

Ifosfamide, MedChemExpress

Product Code. 16461256
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
500 mg
Unit Size:
200mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16461256 200 mg 200mg
16431856 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16461256 Supplier Medchem Express Supplier No. HY17419
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ifosfamide is an alkylating chemotherapeutic agent with activity against a wide range of tumors.

Ifosfamide is an alkylating chemotherapeutic agent with activity against a wide range of tumors.

Specifications

Chemical Name or Material Ifosfamide
Physical Form Solid
CAS 3778-73-2
Color White
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C7H15Cl2N2O2P
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 50 mg/mL (191.50 mM)
SMILES ClCCN1P(OCCC1)(NCCCl)=O
Molecular Weight (g/mol) 261.09
Formula Weight 261.09
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.