Learn More
IFRD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528060
Code : A0
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: Molt-4, Raji. Predicted reactivity: Mouse (94%), Rat (94%), Pig (94%), Chicken (81%), Rhesus Monkey (99%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Tis7, also known as IFRD1, is a 442 amino acid containing membrane associated, non-nuclear intracellular protein belonging to the IFRD family. A regulatory protein expressed in a variety of tissues, Tis7 is known to play an important role in the regulation of gene activity in the proliferative and/or differentiative pathways induced by NGF. Reports suggest that Tis7 may be an autocrine factor that attenuates or amplifies the initial ligand induced signal and might also augment the adaptive response by stimulating the production of differentiated enterocytes. Tis7 acts as a negative regulator of transcriptional activity and represses expression of genes involved in myogenesis, muscle maintenance, and regeneration in a histone deacetylase dependent manner.Specifications
IFRD1 | |
Polyclonal | |
Unconjugated | |
IFRD1 | |
12-O-tetradecanoylphorbol-13-acetate-induced sequence 7; hypothetical protein LOC541283; Ifnl; Ifrd1; interferon related developmental regulator 1; interferon-related developmental regulator 1; IRPR; nerve growth factor-inducible protein PC4; Pc4; pheochromocytoma cell-4; Tis7; TIS7 protein; TPA induced sequence 7; TPA-induced sequence 7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3475 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.62 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
O00458 | |
IFRD1 | |
Recombinant fragment corresponding to a region within amino acids 24 and 409 of Human IFRD1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |