Learn More
IFT20 Rabbit anti-Canine, Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13615-1-AP-20UL
 View more versions of this product
Product Code. 16841914
  • 145.00 € / 20µL
Description
IFT20 is part of intraflagellar transport (IFT) particles involved in ciliary process assembly. IFT20 may play a role in the trafficking of ciliary membrane proteins from the Golgi complex to the cilium.
Specifications
Specifications
IFT20
Polyclonal
Unconjugated
IFT20
hIFT20, IFT20
Rabbit
Antigen Affinity Chromatography
RUO
480620, 55978, 90410
-20°C
Liquid
Immunocytochemistry, Western Blot, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin)
0.22 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q61025, Q8IY31
IFT20
IFT20 Fusion Protein Ag4521
20 μL
Primary
Canine, Human, Mouse