missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IFT81 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554130
This item is not returnable.
View return policy
Description
Immunogen sequence: IKFIMDSLNK EPFRKNYNLI TFDSLEPMQL LQVLSDVLAE IDPKQLVDIR EEMPEQTAKR MLSLLGILKY KPSGNATDMS TFRQGLVIGS KPVIYPVLHW LLQRTNELKK R Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 92%.
Isoform CDV-1 may be involved in cardiac hypertrophy caused by carnitine deficiency. Isoform CDV-1R appears to play a role in development of the testis and spermatogenesis.
Specifications
| IFT81 | |
| Polyclonal | |
| Unconjugated | |
| IFT81 | |
| AW060663; carnitine deficiency-associated gene expressed in ventricle 1; carnitine deficiency-associated protein expressed in ventricle 1; Cdv1; CDV-1; CDV1R; CDV-1R; DV1; Ift81; intraflagellar transport 81; intraflagellar transport 81 homolog; intraflagellar transport protein 81 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 28981 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WYA0 | |
| IFT81 | |
| Recombinant protein corresponding to Human IFT81. Recombinant protein control fragment (Product #RP-92292). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction